Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E2 Tolerances
Brand: Ohaus™ 80780287
863.37 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished, mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
0.200 kg | |
0.008 mg |
Stainless Steel (304) | |
10 mg | |
Protective Case |
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML E2, NVLAP > 10 mg