Learn More
Ohaus™ Stainless Steel Calibration Weight
Adjusted to meet or exceed OIML M1 tolerances
Brand:  Ohaus™ 11117146
373.08 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12773285
  • 386.00 NOK / Each
Only null left
Estimated Shipment: 14-07-2025
Description
  • Meets OIML M1 standards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OIML M1 class
50 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weight > Weight: 50g