Learn More
Usbeck Laborgeraete™ Stainless Steel Bowl
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Capacity (Metric) | 2000 mL |
| Height (Metric) | 115 mm |
| Shape | High |
| Product Type | Bowl |
Product Content Correction
Product Title
Usbeck Laborgeraete™ Stainless Steel Bowl >