Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF55plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ SF55plusB04358
Product Code. 17518325
  • ONLINE EXCLUSIVE PRICE
    1565.00 NOK
    1520.84 NOK / Each
    Save 44.16 NOK
    2% Off
Only null left
Estimated Shipment: 12-11-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF55plus Sterilizer
Bottom Drip Tray
Rim: 15 mm
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for SF55plus, 15 mm > For Use With: SF55plus