Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF30plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ SF30plusB04356
1357.06 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17548235
  • 1395.00 NOK / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF30plus Sterilizer
Bottom Drip Tray
Rim: 15 mm
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for SF30plus, 15 mm > For Use With: SF30plus