Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF260plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand: Memmert™ SF260plusB29722
2772.61 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
SF260plus Sterilizer | |
Bottom Drip Tray | |
Rim: 15 mm | |
Stainless Steel |
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for SF260plus, 15 mm > For Use With: SF260plus