Learn More
Bochem™ Sharp Bent Round Forceps
Finely pointed and curved forceps. Bochem™ Sharp Bent Round Forceps are constructed of stainless steel.
Brand:  Bochem™ 1022R
 View more versions of this product
Product Code. 10213941
  • 123.00 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Stainless steel construction.
  • Durable and resistant.
  • Curved and finely pointed tip.
Specifications
Specifications
Bent, Round
Stainless Steel (18/10)
Forceps
1/PK
145 mm
Forceps
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Sharp Bent Round Forceps > Length: 145mm