Learn More
Socorex™ Stainless Steel Bent Drenching Cannula
Brand:  Socorex™ 376.16102B
 View more versions of this product
Product Code. 10242873
  • 386.00 NOK / Pack of 2
Specifications
Autoclavable
Stainless Steel
102 mm
Drenching Cannula
For safe animal mouth and nose feeding
1.6 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Socorex™ Stainless Steel Bent Drenching Cannula > Diameter: 1.6mm