Learn More
Bochem™ Stainless Steel/ aluminum Ratschet
Brand:  Bochem™ 11088
Product Code. 10078852
  • 590.00 NOK / Each
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Specifications
Ratschet
190 mm
Stainless Steel, Aluminum
1 Ea.
Documents
One moment while we fetch your results.
Product Title
Bochem™ Stainless Steel/ aluminum Ratschet > Length: 190mm