Learn More
Usbeck Laborgeraete™ Stainless Steel Alcohol Burners
59.85 NOK - 276.00 NOK
Specifications
Product Type | Spirit Burner |
---|---|
Material | Stainless Steel |
Capacity (Metric) | 60 mL |
No. per Pack | 1 Ea. |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
10251031
|
Usbeck Laborgeraete™
1500 |
Adjustable Wick and Cap |
276.00 NOK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
10126771
|
Usbeck Laborgeraete™
1501 |
Replacement Wick and Cap |
59.85 NOK
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
Specifications
Spirit Burner | |
Stainless Steel | |
60 mL | |
1 Ea. |
Product Title
Usbeck Laborgeraete™ Stainless Steel Alcohol Burners