Learn More
Sartorius™ Stainless Steel Adapter
Recommended accessories and consumables for use with MD8 Airscan™ Gelatin membrane filter.
Brand: Sartorius™ 17801---001
This item is not returnable.
View return policy
Description
- For installation of the MD8 Airscan™ through the isolator wall
-
Specifications
For gelatin filter units used with MD8 Airscan™ | |
Stainless Steel | |
Stainless steel |