Learn More
Julabo™ Stainless Steel 1-Adaptor
Brand:  Julabo™ 8891008
 View more versions of this product
Product Code. 10752604
  • 1385.00 NOK / Each
Only null left
Estimated Shipment: 30-06-2025
Add to basket
Specifications
Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator
1-Adapter
Stainless Steel
M16 x 1 Male to 1/2 in. BSP Female
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Julabo™ Stainless Steel 1-Adaptor > Connection Type: M16 x 1 Male to 1/2 in. BSP Female