Learn More
Bochem™ Stainless Steel 1:2 Tissue Forceps
Used for dissection
Brand:  Bochem™ 1152
 View more versions of this product
Product Code. 10663911
  • 57.10 NOK / Each
Only null left
Estimated Shipment: 18-09-2025
Description
  • Manufactured from stainless steel
Specifications
Specifications
145 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Stainless Steel 1:2 Tissue Forceps > Length: 145mm