Learn More
Eppendorf™ Stacking and Leveling
Click to view available options
For Use With (Application):
Stack and Level >180 kg
Stack and Level <100 kg
Stack and Level 100 to 180 kg
Unit Size:
Each
Description
- Stacking and leveling Incubators/Shakers
Specifications
Specifications
| For Use With (Equipment) | Incubators and Shakers |
| Service Offering | Stack and Level Equipment |
| For Use With (Application) | Stack and Level 100 to 180 kg |