Learn More
Eppendorf™ Stacking and Leveling
Product Code. 15713695
Stack and Level <100 kg, Each
Click to view available options
For Use With (Application):
Stack and Level >180 kg
Stack and Level <100 kg
Stack and Level 100 to 180 kg
Unit Size:
Each
Product Code. 15713695
Brand: Eppendorf™ 2277000006
Stacking and leveling Incubators/Shakers
  • Stacking and leveling Incubators/Shakers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Incubators and Shakers
Service Offering Stack and Level Equipment
For Use With (Application) Stack and Level <100 kg
Product Content Correction
Product Title
Eppendorf™ Stacking and Leveling >