Learn More
Gilac™ Stackable Reinforced Crate with Handles
Reinforced storage container in food-grade plastic (HDPE). Stackable bin with multi-purpose handles, ideal for handling.
380.23 NOK
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
30245678
|
Gilac™
G154620 |
Gray |
380.23 NOK
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
30245697
|
Gilac™
G154619 |
White |
380.23 NOK
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
Description
- Reinforced 545 x 390 mm storage container in food-grade plastic (HDPE)
- Stackable bin with multi-purpose handles, ideal for handling. Ideal for negative or positive cold storage
- This reinforced, professional-quality bin is designed for intensive use
Specifications
Storage Container |
Product Title
Gilac™ Stackable Reinforced Crate with Handles