missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | STAC |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
16898583
|
Proteintech
11480-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888583
|
Proteintech
11480-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Probably involved in a neuron-specific signal transduction.Spezifikation
STAC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P97306, Q99469 | |
20840, 363152, 6769 | |
STAC Fusion Protein Ag2059 | |
Primary | |
-20°C | |
STAC |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SH3 and cysteine rich domain, STAC, STAC1 | |
Stac | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |