Learn More
Thermo Scientific™ ST8R Cell Culture Centrifuge Package
Meet both clinical and research protocol needs.
Brand: Thermo Scientific™ 50144426
81007.60 NOK valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Includes: Sorvall ST8R, 230V (Mfr PN: 75007204); TX-150 Rotor (Mfr PN: 75005701); TX-150 Roundbucket (set of 4) (Mfr PN: 75005702); Sealing lids (set of 4) (Mfr PN: 75005707); 50mL con skirted and unskirted adapter (set of 4) (Mfr PN: 75005736); 15mL con adapter (set of 4) (Mfr PN: 75005737)
Description
Provides exceptional capacity in a compact design with a smart, simple interface and the flexibility to support numerous applications.
Specifications
ST8R Cell Culture Centrifuge Package | |
Sorvall ST8R, 230V (Mfr PN: 75007204); TX-150 Rotor (Mfr PN: 75005701); TX-150 Roundbucket (set of 4) (Mfr PN: 75005702); Sealing lids (set of 4) (Mfr PN: 75005707); 50mL con skirted and unskirted adapter (set of 4) (Mfr PN: 75005736); 15mL con adapter (set of 4) (Mfr PN: 75005737) |
Promotions
-
Thermo Scientific Sorvall
,
Fisherbrand
,
Thermo Scientific
Make Us an Offer on Centrifuges
Unlock special pricing on top centrifuges. Make us an offer today and upgrade your lab equipment. Limited time only!Promo Code: 21629Expires: 2025-12-31
Days
212Hours
13Minutes
24Seconds
31 -
Multiple Manufacturers
Exclusive Promotion on Equipment and Instruments
Take advantage of our limited-time offer on top-quality equipment and instruments to elevate your research and laboratory capabilities.Promo Code: 24069Expires: 2025-06-30
Days
28Hours
13Minutes
24Seconds
31 -
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31