Learn More
Hettich Lab Technology™ Square Carrier
Square Carrier/Bucket is suitable for Swing-out Rotor 56999-R and accepts a range of adapters for tubes and plates of varying capacity.
Brand: Hettich Lab Technology™ 5628
This item is not returnable.
View return policy
Description
- Open bucket/carrier for adapter inserts
- Suitable for several swing-out rotors and adapters
- Accommodates a variety of centrifuge workloads
Specifications
Swing-Out Rotor 5699-R, Adapter 5227, 5242, 5243, 5249, 5258, 5262, 5263-A, 5264, 5266, 5267, 5268, 5280, 5281, 6337-B, 6338-B, 6339-A, MTP, CP, DWP, MS, QP, Microtest plate (Terasaki), 96-well PCR Plate, PCR Strips | |
Bucket | |
Carrier, Bucket | |
1 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Hettich Lab Technology™ Square Carrier > No. of Places: 1