Learn More
Hecht Karl™ Specific Gravity Bottles
746.00 NOK - 2445.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10798272
|
Hecht Karl™
42571324 |
25 mL |
746.00 NOK
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
10778282
|
Hecht Karl™
42572016 |
10 mL |
2445.00 NOK
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
10796372
|
Hecht Karl™
42572340 |
100 mL |
1845.00 NOK
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
Specifications
Specific Gravity Bottle | |
Specific Gravity Bottle |
Product Title
Hecht Karl™ Specific Gravity Bottles