Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand: 3M™ 7000052909
13301.73 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Fits a wide range of face shapes and sizes- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
10 | |
Red | |
Cup/Molded | |
Particulate Respirator with Nuisance Organic Vapor Relief | |
Yes | |
Electret (polyester, polypropylene, and carbon) | |
Yes | |
Acid Gas | |
Cup | |
No |
EN149:2001 + A1:2009 | |
White | |
Single-use | |
Standard | |
FFP3 | |
Cool-flow Valve, Red Color Coded Straps | |
9936 | |
180/Pk. | |
Non-sterile |