Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Click to view available options
Filter Efficiency:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Color:
Blue
Gray
White
Unit Size:
Pack of 10
Pack of 180
Pack of 20
Beschreibung
Fits a wide range of face shapes and sizes
- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Spezifikation
Spezifikation
Assigned Protection Factor (APF) | 4 |
Certifications/Compliance | EN 149:2001A1:2009, CE. |
Certifications/Compliance (Europe) | EN149:2001 + A1:2009 |
Color | Gray |
Design Type | Cup/Molded |
Disposable | Single-use |
Filter Type | FFP1 |
Product Type | Particulate Respirator with Nuisance Organic Vapor Relief |
Size | Standard |
With Exhalation Valve | No |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
3M™ Aura™ Specialty Respirator, Series 9300 >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur