Learn More
Invitrogen™ Custom DNA Oligos Synthesis Services
Custom Primer
Brand: Invitrogen™ 10629186
1.37 NOK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Store at room temperature. | |
Design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modification | |
Normal or Reversed-phase Chromatography, HPLC or PAGE |
Custom Primer, 25nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification | |
25 nmol | |
Custom Primer |
Product Content Correction
Product Title
Invitrogen™ Custom DNA Oligos Synthesis Services > 25 nmol