Learn More
Invitrogen™ Custom DNA Oligos Synthesis Services
Custom Primer
Brand:  Invitrogen™ 10629053
12.87 NOK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
 View more versions of this product
Product Code. 10022792
  • 17.16 NOK / Each
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 μmol
Custom Primer, 20umole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification
HPLC
Normal or Reversed-phase Chromatography, HPLC or PAGE
Custom Primer
Design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modification
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ Custom DNA Oligos Synthesis Services > 1μmol