Learn More
SPDP, MedChemExpress Product Code.: 16471516
SPDP, MedChemExpress
Product Code. 16471516
500 mg, 500mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Product Code. 16471516
Brand: Medchem Express HY100216
MedChemExpress SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
Specifications
Chemical Name or Material SPDP
CAS 68181-17-9
Color White
Purity Grade Notes Research
Molecular Formula C12H12N2O4S2
Quantity 500 mg
Synonym SPDP Crosslinker
Solubility Information DMSO : 155 mg/mL (496.22 mM; Need ultrasonic)
SMILES O=C(ON1C(CCC1=O)=O)CCSSC2=NC=CC=C2
Molecular Weight (g/mol) 312.36
Formula Weight 312.36
Percent Purity 97.0%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SPDP, MedChemExpress