Learn More
Eppendorf™ Spare Gaskets
For use with Eppendorf liquid handling products
Brand: Eppendorf™ 5810713005
880.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory for Eppendorf caps and rotors
- Spare sealing and replacement gasket
Specifications
Spare Sealing | |
250mL Aerosol-Tight Cap | |
Replacement Gasket | |
Pack of 4 |
Product Content Correction
Product Title
Eppendorf™ Spare Gaskets > For 250mL Aerosol-Tight Cap