Learn More
Eppendorf™ Spare Bottom Disc for Bucket
Rubber spare bottom disc for use with Rotor A-4-38 Bucket.
Brand: Eppendorf™ 5702764019
670.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rubber
- For 16 x 125 mm bucket
- For rotor A-4-38
- Accessory
Specifications
Bucket for Rotor A-4-38 | |
Adapters, Buckets and Accessories | |
8/Pk. |