Learn More
Socorex™ Spare Kit Bag, Socorex™ 187 Syringe
Replace valve assembly parts for the Socorex™ 187 Tube Filling syringe if necessary. Socorex™ Spare Kit Bag, Socorex™ 187 Syringe includes all the spare parts needed for repair/replacement of your Socorex™ 187 Syringe valve.
Brand:  Socorex™ 1.187.7202
 View more versions of this product
Product Code. 10580943
  • 117.00 NOK / Each
Explore more special offers
Description
Description
  • Spare parts for Socorex™ 187 Tube Feeding Syringe models based on syringe volume.
  • Each bag includes the following:
    • 2/ea. springs.
    • 2/ea. balls.
    • 2/ea. valve washers.
    • 1/ea. barrel washer.
Specifications
Specifications
2/ea. springs, 2/ea. balls, 2/ea. valve washers, one barrel washer.
Spare Kit Bag
2 mL
Socorex 187 Tube Feeding Syringe, 2 mL
Documents
One moment while we fetch your results.
Product Title
Socorex™ Spare Kit Bag, Socorex™ 187 Syringe > Volume: 2mL