missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG4L Rabbit anti-Human, Mouse, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-17495-20UL
This item is not returnable.
View return policy
Description
SPAG4L Polyclonal antibody specifically detects SPAG4L in Human, Mouse samples. It is validated for FC (Intra)Specifications
SPAG4L | |
Polyclonal | |
CoraLite 594 | |
BC005127 | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Rabbit | |
20 μL | |
140732 | |
Store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction