Learn More
3M™ Solus™ Safety Glasses
3M™ Solus™ Safety Glasses are stylish and lightweight for improved comfort and wearer acceptance.
1997.84 NOK - 2855.00 NOK
Specifications
Certifications/Compliance | CE Approved, EN 166: 2001 |
---|---|
Product Type | Safety glasses |
Includes | Microfiber storage bag |
Lens Material | Polycarbonate |
Product Code | Brand | Lens Tint | Lens Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Tint | Lens Coating | Price | Quantity & Availability | |||||
15719677
|
3M™
7000032474 |
Bronze | Anti-fog, Scratch-resistant | N/A | ||||||
15729677
|
3M™
7000032475 |
Amber | Anti-fog, Scratch-resistant | N/A | ||||||
15739677
|
3M™
7000032476 |
Indoor/Outdoor, Mirror | Scratch-resistant | N/A | ||||||
15739807
|
3M™
7000061911 |
Clear | Scratch-resistant | N/A | ||||||
15749677
|
3M™
7000032478 |
Blue, Mirror | Scratch-resistant | N/A | ||||||
15749807
|
3M™
7000061912 |
Red, Mirror | Scratch-resistant | N/A | ||||||
15759807
|
3M™
7000061914 |
Clear | Anti-fog, Scratch-resistant | N/A | ||||||
Description
- Lens curve has been calculated to optimize the fitting, field of vision and coverage of the eyes
- Extremely stylish and lightweight glasses
- Soft inner padding of the temple, leading to increased comfort
- Supplied with a microfiber storage bag for cleaning and protection
- Anti-scratch and anti-fog coating for increased durability and improved vision
Specifications
CE Approved, EN 166: 2001 | |
Safety glasses | |
Microfiber storage bag | |
Polycarbonate |
Product Title
3M™ Solus™ Safety Glasses