Learn More
3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
251.00 NOK - 7375.00 NOK
Specifications
Lens Coating | Anti-fog |
---|---|
For Use With (Application) | Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
Frame Type | Half Frame |
Product Code | Brand | Product Type | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Frame Color | Lens Tint | Price | Quantity & Availability | ||||
15767717
|
3M™
7100080258 |
Safety Glasses | Blue, Black | Clear |
251.00 NOK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||
15566565
|
3M™
7100080184 |
Safety Glasses Kit | Blue, Black | Clear |
7080.00 NOK
Pack of 20 |
In Stock
Log in to see stock. ![]() |
||||
15546565
|
3M™
7100080185 |
Safety Glasses | Blue, Black | Gray |
5045.00 NOK
Pack of 20 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
15556565
|
3M™
7100080187 |
Safety Glasses | Blue, Black | Amber |
5010.00 NOK
Pack of 20 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
15707717
|
3M™
7100078882 |
Safety Glasses | Green, Black | Clear |
251.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
15506575
|
3M™
7100078881 |
Safety Glasses Kit | Green, Black | Clear |
7375.00 NOK
Pack of 20 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
15586565
|
3M™
7100078883 |
Safety Glasses | Green, Black | Gray |
5070.00 NOK
Pack of 20 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
15596565
|
3M™
7100080186 |
Safety Glasses | Green, Black | Amber |
4820.00 NOK
Pack of 20 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
Description
- Stylish and modern look slim frame with colour accented soft temple components
- Strong polycarbonate lens for impact resistance
- 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
- Lightweight with soft cushioning on nose bridge
- Inner padding on temples for increased comfort and stability when worn
- Meets K&N marking requirements of EN 166
- Frame marking: 3M EN166 FT CE
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Anti-fog | |
Polycarbonate | |
Half Frame |
Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation | |
Polycarbonate |