Learn More
Thermo Scientific™ Solid Door Option with Inner Glass Door for Large Chamber
Protection for your light sensitive samples
Brand:  Thermo Scientific™ 1900667
Additional Details : Weight : 0.01000kg
Product Code. 16491125
  • 31200.71 NOK / Each
Estimated Shipment: 03-10-2024
Description
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifications
Specifications
Solid door with inner glass door for large chamber, replaces standard outer glass door, factory installed
LED
Solid door with inner glass door for large chamber
Factory installed solid door with inner glass door
order with chamber models 3920, 3940, 3948, 3960, 3962
Accessory, Solid Door
incubation, light sensitive applications