Learn More
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For Hcp50
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ HCP50FDAQ1
Product Code. 16394877
  • 32935.00 NOK / Each
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106
Software
Product Suggestions
Product Suggestions
Videos
Videos
  • Memmert AtmoCONTROL (EN)
  • Memmert ControlCOCKPIT (EN)
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For Hcp50 >