Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black/Yellow
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
Brand:  Notrax 411S0323BY
 View more versions of this product
Product Code. 17722088
  • 343.00 NOK / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Description
  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Specifications
Black/Yellow
91 cm
Industrial and Commercial Matting
36 in.
Microcellular Vinyl
60 cm