missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiocyanate solution 0.01mol/L, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
CAS | 7732-18-5 |
Quantity | 1000 mL |
Synonym | dihydrogen oxide, dihydrogen monoxide |
Concentration or Composition (by Analyte or Components) | 0.01M (0.01N) |
Packaging | Bottle |
Chemical Name or Material | Sodium Thiocyanate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Thiocyanate solution 0.01mol/L, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction