Learn More
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 140.0°C to 142.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: reacts |
Formula Weight | 150.04 |
Percent Purity | 97% |
Grade | Pure |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0468712 | Certificate of Analysis | 31/01/2025 | 363880050,363880010 |
A0448502 | Certificate of Analysis | 31/01/2025 | 363880050,363880010 |
A0466550 | Certificate of Analysis | 31/10/2024 | 363880050,363880010 |
A0464382 | Certificate of Analysis | 15/08/2024 | 363880050,363880010 |
A0452444 | Certificate of Analysis | 24/04/2024 | 363880050,363880010 |
Safety and Handling

- Sodium tetraethylborate
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P231+P232-Handle under inert gas. Protect from moisture.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P335-Brush off loose particles from skin.
- P370+P378-In case of fire: Use to extinguish.
- P422-Store contents under .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.