missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, Reagent Grade, ≥99.5%, Honeywell™

Reagent Grade,≥99.5%

401.50 NOK - 2407.88 NOK

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15637710
SDS View Documents
Honeywell Research Chemicals
B9876-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15627710
SDS View Documents
Honeywell Research Chemicals
B9876-1KG
1 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15647710
SDS View Documents
Honeywell Research Chemicals
B9876-5KG
5 kg Drum with inliner This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1303-96-4
MFCD00149193

Specifications

1575°C
1303-96-4
9 to 10 (at 20°C)
Na2B4O7 · 10H2O
NONH for all modes of transport
≥99.5%
1.73g/mL at 25°C
62°C
Colorless
Solid
MFCD00149193
381.37g/mol
Odorless
Sodium tetraborate decahydrate
Videos
Safety and Handling

Safety and Handling

P201-P280-P308 + P313

missing translation for 'rtecsNumber' : 215-540-4

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Tetraborate Decahydrate, Reagent Grade, ≥99.5%, Honeywell™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.