missing translation for 'onlineSavingsMsg'
Learn More
Sodium Sulfide Hydrate, Honeywell™ Product Code.: 15691440

Sodium Sulfide Hydrate, Honeywell™

Product Code. 15691440 Shop All Honeywell Research Chemicals Products
250 g, Glass Bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15691440

Brand: Honeywell Research Chemicals 14738250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

for gravimetric analysis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]

Specifications

Color Colorless
Quantity 250 g
UN Number UN1849
IUPAC Name disodium;sulfanide;hydrate
Formula Weight 78.04g/mol (anhydrous basis)
Percent Purity 59 to 65%
Packaging Glass Bottle
Appearance Flakes
Chemical Name or Material Sodium Sulfide Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfide Hydrate, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.