missing translation for 'onlineSavingsMsg'
Learn More
Sodium Sulfate Anhydrous, Pharmpur™, Scharlab Product Code.: 11338564

Sodium Sulfate Anhydrous, Pharmpur™, Scharlab

Product Code. 11338564
5 kg, 5kg
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
This item is not returnable. View return policy

Product Code. 11338564

Brand: Scharlab SO0665005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, and in organic solvents drying.

  • Synonyms: Sulfuric acid sodium salt
  • Molecular Formula: Na2SO4
  • Molar Mass: 142.04g/mol
  • CAS: 7757-82-6
  • EIN ECS No.: 231-820-9

Specifications

Boiling Point >890°C (decomposition)
Melting Point 888°C
CAS 7757-82-6
Physical Form Powder
Quantity 5 kg
Molecular Formula Na2O4S
MDL Number MFCD00003504
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium sulfate
Molecular Weight (g/mol) 142.04
PubChem CID 24436
ChEBI CHEBI:32149
Formula Weight 142.04g/mol
Percent Purity 98.5 to 101% (on dried sample)
Grade Pharmpur
Identification Passes Test
Loss on Drying 0.5% max (130°C)
Appearance Clear, Colorless
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfate Anhydrous, Pharmpur™, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.