missing translation for 'onlineSavingsMsg'
Learn More
Sodium Standard Solution, 1000ÎĽg/mL, Spectro ECON, Chem-Lab Product Code.: 11968504

Sodium Standard Solution, 1000ÎĽg/mL, Spectro ECON, Chem-Lab

Product Code. 11968504
Water
Click to view available options
Solvent or Matrix:
Nitric Acid
Water
This item is not returnable. View return policy

Product Code. 11968504

Brand: Chem Lab CL01.1405.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a reference standard in spectroscopy and other laboratory applications. Sodium Standard Solution, 1000ÎĽg/mL, Spectro ECON, Chem-Lab is available in a matrix of water or nitric acid to meet the needs of different analytical processes.

This sodium standard solution contains a precise concentration of 990–1010μg/mL of Na1+ (±0.2%). It is used to verify the accuracy of sodium determinations and to calibrate equipment used in those determinations.

Chemical Identifiers

CAS 7647-14-5, 7732-18-5

Specifications

Concentration or Composition Notes 2.542g NaCl per 1L H2O
Density 1.00g/mL
Physical Form Liquid
Molecular Formula ClNa
Concentration 1000 μg/mL of Na1+ (±0.2%)
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Solvent or Matrix Water
Molecular Weight (g/mol) 58.44
PubChem CID 5234
Quantity 500 mL
ChEBI CHEBI:26710
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Standard Solution, 1000ÎĽg/mL, Spectro ECON, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.