missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxodisulfate, EssentQ, Scharlab

Product Code. 11388554
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7775-27-1
Quantity 1 kg
Molecular Formula Na2O8S2
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name disodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 238.09
Grade EssentQ
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium peroxodisulfate, extra pure
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'