missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate Monohydrate, 98%, ACS Reagent, Honeywell Fluka™

ACS Reagent, 98%

333.96 NOK - 1690.36 NOK

Chemical Identifiers

CAS 7791-07-3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15693190
SDS View Documents
Honeywell-Fluka
310514-100G
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15603200
SDS View Documents
Honeywell-Fluka
310514-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P220

H271-H302

missing translation for 'dotInformation' : 5.1/PGII

missing translation for 'einecsNumber' : 231-511-9

missing translation for 'rtecsNumber' : SC9850000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Perchlorate Monohydrate, 98%, ACS Reagent, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.