missing translation for 'onlineSavingsMsg'
Learn More
Sodium Pentacyanoammine Ferroate, MP Biomedicals Product Code.: 11407734

Sodium Pentacyanoammine Ferroate, MP Biomedicals

Product Code. 11407734 Shop All MP Biomedicals Products
10 g, 10g
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11407734

Brand: MP Biomedicals 0520302510

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 80789-79-3
Molecular Formula C5H17FeN7Na2O5
Quantity 10 g
Synonym sodium pentacyanoammine ferroate, ammonium dipotassium aminiopentacyanoironbis ylium pentahydrate
InChI Key OTKYGVKYEZPNBM-UHFFFAOYSA-O
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].N.O.O.O.O.O.[Na+].[Na+].[Fe+2]
IUPAC Name azanium;disodium;azane;iron(2+);pentacyanide;pentahydrate
Molecular Weight (g/mol) 357.06
PubChem CID 25022167
Formula Weight 266.99
Chemical Name or Material Sodium Pentacyanoammine Ferroate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Pentacyanoammine Ferroate, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.