missing translation for 'onlineSavingsMsg'
Learn More
Sodium n-octyl sulfate, 99% Product Code.: 11449363

Sodium n-octyl sulfate, 99%

Product Code. p-7081707
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11449363

Brand: Thermo Scientific Alfa Aesar A11786.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-31-4 | C8H17NaO4S | 232.27 g/mol

Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H17NaO4S
Molecular Weight (g/mol) 232.27
MDL Number MFCD00007470
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼195°C (decomposition)
Quantity 1 g
Beilstein 3658635
Solubility Information Soluble in water.
Formula Weight 232.28
Percent Purity 99%
Chemical Name or Material Sodium n-octyl sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberX08K001 Certificate TypeCertificate of Analysis Date18/10/2024 Product CodeA11786.06,A11786.03,A11786.14
Lot Number10251485 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA11786.06,A11786.03,A11786.14
Lot NumberM24K008 Certificate TypeCertificate of Analysis Date31/01/2024 Product CodeA11786.06,A11786.03,A11786.14
Lot Number10248928 Certificate TypeCertificate of Analysis Date26/01/2024 Product CodeA11786.06,A11786.03,A11786.14
Lot NumberQ20H041 Certificate TypeCertificate of Analysis Date05/01/2023 Product CodeA11786.06,A11786.03,A11786.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium n-octyl sulfate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.