missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, 99.99% Trace Metals Basis, Honeywell™
99.99% trace metals basis
1064.36 NOK - 3314.42 NOK
Chemical Identifiers
CAS | 10102-40-6 |
---|---|
Molecular Formula | MoNa2O4 |
Molecular Weight (g/mol) | 205.93 |
MDL Number | MFCD00149170 |
InChI Key | TVXXNOYZHKPKGW-UHFFFAOYSA-N |
Synonym | sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate |
PubChem CID | 16211258 |
ChEBI | CHEBI:75213 |
IUPAC Name | disodium dioxomolybdenumbis(olate) |
SMILES | [Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
10102-40-6 | |
205.93 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
16211258 | |
disodium dioxomolybdenumbis(olate) |
MoNa2O4 | |
MFCD00149170 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
CHEBI:75213 | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Specifications
100°C | |
White | |
Liquid | |
Na2MoO4 · 2H2O | |
NONH for all modes of transport | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
disodium dioxomolybdenumbis(olate) | |
16211258 | |
241.95g/mol | |
Odorless | |
Sodium molybdate dihydrate |
10102-40-6 | |
7 to 10 (at 20°C) | |
MoNa2O4 | |
MFCD00149170 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
205.93 | |
CHEBI:75213 | |
99.99% (trace metals basis) | |
2.37g/mL at 25°C |
Safety and Handling
missing translation for 'rtecsNumber' : 231-551-7
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Molybdate Dihydrate, 99.99% Trace Metals Basis, Honeywell™