missing translation for 'onlineSavingsMsg'
Learn More

Sodium Meta-Silicate, 9-Hydrate, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

8535.00 NOK

Chemical Identifiers

CAS 13517-24-3
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15558184
View Documents Promotion Details
Avantor J.T.Baker
3868-01
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

13517-24-3
284.197
sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
disodium;dioxido(oxo)silane;nonahydrate
H18Na2O12Si
PHIQPXBZDGYJOG-UHFFFAOYSA-N
61639
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

13517-24-3
sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
284.197
284.2
Sodium Meta-Silicate, 9-Hydrate
H18Na2O12Si
PHIQPXBZDGYJOG-UHFFFAOYSA-N
disodium;dioxido(oxo)silane;nonahydrate
61639
BAKER ANALYZED™ Reagent
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium meta-Silicate, 9-Hydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium meta-Silicate, 9-Hydrate

SDS
Documents

Documents

Product Certifications