missing translation for 'onlineSavingsMsg'
Learn More
Sodium mercury amalgam, ca. 20% sodium Product Code.: 10751714

Sodium mercury amalgam, ca. 20% sodium

Product Code. p-3593519 Shop All Thermo Scientific Chemicals Products
10 g, Glass bottle, 10g
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10751714

Brand: Thermo Scientific Acros 388460100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11110-52-4 | HgNa | 223.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11110-52-4
Molecular Formula HgNa
Molecular Weight (g/mol) 223.582
MDL Number MFCD00210362
InChI Key MJGFBOZCAJSGQW-UHFFFAOYSA-N
Synonym sodium amalgam, sodium mercury amalgam, sodium mercury, sodium-amalgam, mercury sodium, sodium mercury amalgam, beads, na 10 %, sodium mercury amalgam, beads trace metals basis, na 5 %, sodium mercury amalgam, beads trace metals basis, na 20 %
PubChem CID 9837419
IUPAC Name mercury;sodium
SMILES [Na].[Hg]

Specifications

Color Gray
Quantity 10 g
Merck Index 15, 8713
Packaging Glass bottle
Form Pieces
Chemical Name or Material Sodium mercury amalgam

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0459629 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code388460500,388460100
Lot NumberA0468449 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code388460500,388460100
Lot NumberA0450363 Certificate TypeCertificate of Analysis Date22/02/2024 Product Code388460500,388460100
Lot NumberA0459629 Certificate TypeCertificate of Analysis Date26/01/2024 Product Code388460500,388460100
Lot NumberA0396062 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code388460500,388460100
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium mercury amalgam, ca. 20% sodium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H360-May damage fertility or the unborn child.
  • H330-Fatal if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium mercury amalgam, ca. 20% sodium >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.