missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, 5.0M, Chem-Lab
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
Unit Size:
1 L
Description
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
Specifications
pH | 13 |
Color | Colorless |
Boiling Point | 100°C |
CAS | 1310-73-2 |
Molecular Formula | HNaO |
Concentration | 5 mol/L |
Concentration or Composition Notes | 200g NaOH per 1L H2O |
Density | 1.18g/mL |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Water Soluble |
Show More |
Safety and Handling

Product Identifier
- Sodium hydroxide 5 mol/l
- Danger
- Skin corrosion/irritation Category 1 A
- Substances and mixtures corrosive to metals Category 1
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sodium hydroxide, pellets a.r.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Solution, 5.0M, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction