missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide Solution, 40% Weight, Chem-Lab Product Code.: 11954004
compliance-icons

Sodium Hydroxide Solution, 40% Weight, Chem-Lab

Product Code. 11954004
5 L, 5L, 5 L
Click to view available options
Quantity:
5 L
Packaging:
5L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 11954004

Brand: Chem Lab CL02.1410.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in laboratory applications. Sodium Hydroxide Solution, 40% Weight, Chem-Lab is a strong base and is highly caustic. It is used in titration and to increase alkalinity, to neutralize acids and to make sodium salts. One liter of the solution contains 572g of NaOH.

Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.

Specifications

Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 40% by weight
Density 1.43g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Concentration or Composition (by Analyte or Components) 572g NaOH per 1L H2O
Odor Odourless
Packaging 5L
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 40 weight % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide Solution, 40% Weight, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.