missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide, 0.1M, Chem Lab™ Product Code.: 11963213
compliance-icons

Sodium Hydroxide, 0.1M, Chem Lab™

Product Code. 11963213
10 L, 10 L
Click to view available options
Quantity:
10 L
Unit Size:
10 L
This item is not returnable. View return policy

Product Code. 11963213

Brand: Chem Lab CL05.1411.9010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH >11
Color Colorless
Concentration 4g NaOH/l H2O = 0.1 N (±0.0002/20°C)
Density 1.01g/mL
Solubility Information Soluble in Water
Quantity 10 L
Odor Odorless
Physical Form Liquid
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, 0.1M, Chem Lab™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.